Concanavalin A (Con A) Conjugates
Invitrogen™

Concanavalin A (Con A) Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateColorExcitation/EmissionProtein Subtype
C21401Alexa Fluor 488Green495/519 nmConcanavalin A, succinylated
C11254Alexa Fluor 350Blue346/442 nmConcanavalin A (Con A)
C11252Alexa Fluor 488Green495/519 nmConcanavalin A (Con A)
C11253Alexa Fluor 594Red590/617 nmConcanavalin A (Con A)
C21402Alexa Fluor 633Deep Red632/647 nmConcanavalin A (Con A)
C21421Alexa Fluor 647Deep Red650/668 nmConcanavalin A (Con A)
C56126Alexa Fluor Plus 405Violet405/450 nmConcanavalin A (Con A)
C56127Alexa Fluor Plus 750Near-infrared750/777 nmConcanavalin A (Con A)
C825Texas RedRed595/615 nmConcanavalin A (Con A)
C860TetramethylrhodamineOrange555/580 nmConcanavalin A (Con A)
Catalog number C21401
Price (USD)
235.65
Online exclusive
256.00
Save 20.35
Each
Conjugate:
Alexa Fluor 488
Color:
Green
Excitation/Emission:
495/519 nm
Protein Subtype:
Concanavalin A, succinylated
Price (USD)
235.65
Online exclusive
256.00
Save 20.35
Each
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC). 

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Cross Reactivityα-man, α-glc
Excitation/Emission495/519 nm
Expression SystemNatural
Label TypeAlexa Fluor Dyes
Ligand TypeLectin
Product LineAlexa Fluor
Protein FamilyLectins
Protein SubtypeConcanavalin A, succinylated
Protein TagNone
Purity or Quality GradeImaging Quality
Quantity5 mg
Research CategoryImaging
Shipping ConditionRoom Temperature
SourceCanavlia ensiformis
ConjugateAlexa Fluor 488
For Use With (Application)Immunocytochemistry, Immunohistochemistry, Immunofluorescence
FormLyophilized
RecombinantNative
SpeciesC. ensiformis
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (1)

Citations & References
Abstract
Localization and functionality of the inflammasome in neutrophils.
Authors:Bakele M, Joos M, Burdi S, Allgaier N, Pöschel S, Fehrenbacher B, Schaller M, Marcos V, Kümmerle-Deschner J, Rieber N, Borregaard N, Yazdi A, Hector A, Hartl D,
Journal:
PubMed ID:24398679
Neutrophils represent the major fraction of circulating immune cells and are rapidly recruited to sites of infection and inflammation. The inflammasome is a multiprotein complex that regulates the generation of IL-1 family proteins. The precise subcellular localization and functionality of the inflammasome in human neutrophils are poorly defined. Here we ... More