Concanavalin A (Con A) Conjugates
Invitrogen™
Concanavalin A (Con A) Conjugates
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호콘주게이트색상여기/방출단백질 아형
C21402Alexa Fluor 633Deep Red632/647 nmConcanavalin A (Con A)
C11254Alexa Fluor 350Blue346/442 nmConcanavalin A (Con A)
C11252Alexa Fluor 488Green495/519 nmConcanavalin A (Con A)
C21401Alexa Fluor 488Green495/519 nmConcanavalin A, succinylated
C11253Alexa Fluor 594Red590/617 nmConcanavalin A (Con A)
C21421Alexa Fluor 647Deep Red650/668 nmConcanavalin A (Con A)
C56126Alexa Fluor Plus 405Violet405/450 nmConcanavalin A (Con A)
C56127Alexa Fluor Plus 750Near-infrared750/777 nmConcanavalin A (Con A)
C825Texas RedRed595/615 nmConcanavalin A (Con A)
C860TetramethylrhodamineOrange555/580 nmConcanavalin A (Con A)
카탈로그 번호 C21402
제품 가격(KRW)
330,000
온라인 행사
Ends: 31-Dec-2026
388,000
할인액 58,000
Each
콘주게이트:
Alexa Fluor 633
색상:
Deep Red
여기/방출:
632/647 nm
단백질 아형:
Concanavalin A (Con A)
제품 가격(KRW)
330,000
온라인 행사
Ends: 31-Dec-2026
388,000
할인액 58,000
Each
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC). 
For Research Use Only. Not for use in diagnostic procedures.
사양
색상Deep Red
교차 반응성α-man, α-glc
여기/방출632/647 nm
발현 시스템Natural
라벨 유형Alexa Fluor Dyes
리간드 유형Lectin
제품라인Alexa Fluor
단백질군Lectins
단백질 아형Concanavalin A (Con A)
단백질 태그None
순도 또는 품질 등급Imaging Quality
수량5 mg
연구 카테고리Imaging
배송 조건Room Temperature
소스Canavlia ensiformis
콘주게이트Alexa Fluor 633
용도(애플리케이션)Immunocytochemistry, Immunohistochemistry, Immunofluorescence
형태Lyophilized
재조합Native
종C. ensiformis
Unit SizeEach
구성 및 보관
Store in freezer (-5 to -30°C) and protect from light.
인용 및 참조 문헌 (4)
인용 및 참조 문헌
Abstract
A protein interaction map for cell polarity development.
Authors:Drees BL, Sundin B, Brazeau E, Caviston JP, Chen GC, Guo W, Kozminski KG, Lau MW, Moskow JJ, Tong A, Schenkman LR, McKenzie A, Brennwald P, Longtine M, Bi E, Chan C, Novick P, Boone C, Pringle JR, Davis TN, Fields S, Drubin DG
Journal:J Cell Biol
PubMed ID:11489916
'Many genes required for cell polarity development in budding yeast have been identified and arranged into a functional hierarchy. Core elements of the hierarchy are widely conserved, underlying cell polarity development in diverse eukaryotes. To enumerate more fully the protein-protein interactions that mediate cell polarity development, and to uncover novel ... More
Essential function of Drosophila Sec6 in apical exocytosis of epithelial photoreceptor cells.
Authors:Beronja S, Laprise P, Papoulas O, Pellikka M, Sisson J, Tepass U
Journal:J Cell Biol
PubMed ID:15897260
Polarized exocytosis plays a major role in development and cell differentiation but the mechanisms that target exocytosis to specific membrane domains in animal cells are still poorly understood. We characterized Drosophila Sec6, a component of the exocyst complex that is believed to tether secretory vesicles to specific plasma membrane sites. ... More
The plane of first cleavage is not related to the distribution of sperm components in the mouse.
Authors:Davies TJ, Gardner RL
Journal:Hum Reprod
PubMed ID:12202427
BACKGROUND: Marking experiments using phytohaemagglutinin (PHA) and Concanavalin A (ConA) have suggested that the first cleavage plane is related to the point of sperm entry. Because of concerns about the specificity of lectin binding, the distribution of sperm components has been investigated directly. METHODS AND RESULTS: The sperm tail could ... More
Local Adaptation of Bitter Taste and Ecological Speciation in a Wild Mammal.
Authors:
Journal:Mol Biol Evol
PubMed ID:34240186