Calcium Green™-1, AM, cell permeant
Invitrogen™

Calcium Green™-1, AM, cell permeant

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
C3011MP500 μg
C3012
also known as C-3012
10 x 50 μg
Catalog number C3011MP
Price (USD)
412.00
Each
Quantity:
500 μg
Request bulk or custom format
Price (USD)
412.00
Each

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. The fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry.

Calcium Indicator (AM Ester) Specifications:

  • Label (Ex/Em): Calcium Green™ -1 (506/531 nm)
  • Fluorescence intensity increase upon binding Ca2+ : ∼14 fold
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Spectral Characteristics of Molecular Probes™ Calcium Indicators

These probes are excited by visible light, and because the energy required for excitation is low, the potential for cellular photodamage is reduced. Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem.

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios, for example dextran versions for reduced leakage and compartmentalization and BAPTA conjugates for detecting high-amplitude calcium transients. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeCalcium Green
Emission Wavelength531 nm
Excitation Wavelength506 nm
Quantity500 μg
Shipping ConditionRoom Temperature
Emission Wavelength Range525 to 540 nm
Excitation Wavelength Range495 to 510 nm
For Use With (Application)Calcium Indicator
For Use With (Equipment)Fluorescence Microscope
Product LineCalcium Green
Product TypeCalcium Indicator
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (90)

Citations & References
Abstract
Mechanism of collagen activation in human platelets.
Authors:Roberts DE, McNicol A, Bose R
Journal:J Biol Chem
PubMed ID:14981087
The mechanism of collagen-induced human platelet activation was examined using Ca2+, Na+, and the pH-sensitive fluorescent dyes calcium green/fura red, sodium-binding benzofuran isophthalate, and 2',7'-bis(2-carboxyethyl)-5(6)-carboxyfluorescein. Administration of a moderate dose of collagen (10 microg/ml) to human platelets resulted in an increase in [Ca2+](i) and platelet aggregation. The majority of this ... More
The growth-related gene product beta induces sphingomyelin hydrolysis and activation of c-Jun N-terminal kinase in rat cerebellar granule neurones.
Authors:Limatola C, Mileo AM, Giovannelli A, Vacca F, Ciotti MT, Mercanti D, Santoni A, Eusebi F
Journal:J Biol Chem
PubMed ID:10593952
'The growth-related gene product beta (GRObeta) is a small chemoattractant cytokine that belongs to the CXC chemokine family, and GRObeta receptors are expressed in the brain, including the cerebellum. We demonstrate that rat cerebellar granule neurones express the GRObeta receptor CXCR2. We also show that, in addition to the known ... More
Intracellular astrocyte calcium waves in situ increase the frequency of spontaneous AMPA receptor currents in CA1 pyramidal neurons.
Authors:Fiacco TA, McCarthy KD
Journal:J Neurosci
PubMed ID:14736858
'Spontaneous neurotransmitter release and activation of group I metabotropic glutamate receptors (mGluRs) each play a role in the plasticity of neuronal synapses. Astrocytes may contribute to short- and long-term synaptic changes by signaling to neurons via these processes. Spontaneous whole-cell AMPA receptor (AMPAR) currents were recorded in CA1 pyramidal cells ... More
Control of IP(3)-mediated Ca2+ puffs in Xenopus laevis oocytes by the Ca2+-binding protein parvalbumin.
Authors:John LM, Mosquera-Caro M, Camacho P, Lechleiter JD
Journal:J Physiol
PubMed ID:11507154
'1. Elementary events of Ca2+ release (Ca2+ puffs) can be elicited from discrete clusters of inositol 1,4,5 trisphosphate receptors (IP(3)Rs) at low concentrations of IP(3). Ca(2+) puffs have rarely been observed unless elicited by either hormone treatment or introduction of IP(3) into the cell. However, cells appear to have sufficient ... More
Inositol 1,4,5-trisphosphate and ryanodine receptor distributions and patterns of acetylcholine- and caffeine-induced calcium release in cultured mouse hippocampal neurons.
Authors:Seymour-Laurent KJ, Barish ME
Journal:J Neurosci
PubMed ID:7722617
'The distributions of inositol 1,4,5-trisphosphate and ryanodine receptors (InsP3R and RyR) and the patterns of increase in intracellular calcium ion concentration ([Ca2+]i) elicited by their activation were compared in cultured hippocampal neurons. InsP3R and RyR were labeled using specific antibodies and formed small aggregations in the somata and dendrites of ... More