CultureWell™ Chambered Coverglass for Cell Culture, 16 wells per coverglass
Invitrogen™
CultureWell™ Chambered Coverglass for Cell Culture, 16 wells per coverglass
The CultureWell™ removable chambered coverglass for cell culture is ideal for cell culture and fluorescence imaging applications. The chambered coverglass—providedRead more
Have Questions?
Catalog NumberQuantity
C370001 set
Catalog number C37000
Price (USD)
323.65
Online exclusive
348.00
Save 24.35
Each
Quantity:
1 set
Price (USD)
323.65
Online exclusive
348.00
Save 24.35
Each
The CultureWell™ removable chambered coverglass for cell culture is ideal for cell culture and fluorescence imaging applications. The chambered coverglass—provided sterile and ready to use—contains 16 wells that can each hold up to 250 μL, allowing cells to be cultured in a number of different conditions on a single slide. When the cells are ready to be imaged, coverglass removal is made easy by the use of a simple tool (Cat. No. C37010) that separates the parts without the need for excessive force, eliminating the risk of coverglass breakage.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 set
Shipping ConditionRoom Temperature
Culture Area0.4 cm2
SterilitySterile
Surface TreatmentCell Culture
TypeCoverglass
Volume (Metric) Working250 μL
Wells16
Unit SizeEach
Contents & Storage
Store at room temperature.