Concanavalin A (Con A) Conjugates
Invitrogen™

Concanavalin A (Con A) Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateColorExcitation/EmissionProtein Subtype
C825Texas RedRed595/615 nmConcanavalin A (Con A)
C11254Alexa Fluor 350Blue346/442 nmConcanavalin A (Con A)
C11252Alexa Fluor 488Green495/519 nmConcanavalin A (Con A)
C21401Alexa Fluor 488Green495/519 nmConcanavalin A, succinylated
C11253Alexa Fluor 594Red590/617 nmConcanavalin A (Con A)
C21402Alexa Fluor 633Deep Red632/647 nmConcanavalin A (Con A)
C21421Alexa Fluor 647Deep Red650/668 nmConcanavalin A (Con A)
C56126Alexa Fluor Plus 405Violet405/450 nmConcanavalin A (Con A)
C56127Alexa Fluor Plus 750Near-infrared750/777 nmConcanavalin A (Con A)
C860TetramethylrhodamineOrange555/580 nmConcanavalin A (Con A)
Catalog number C825
Price (USD)
235.65
Online exclusive
256.00
Save 20.35
Each
Conjugate:
Texas Red
Color:
Red
Excitation/Emission:
595/615 nm
Protein Subtype:
Concanavalin A (Con A)
Price (USD)
235.65
Online exclusive
256.00
Save 20.35
Each
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC). 

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorRed
Cross Reactivityα-man, α-glc
Excitation/Emission595/615 nm
Expression SystemNatural
Label TypeClassic Dyes
Ligand TypeLectin
Product LineTexas Red
Protein FamilyLectins
Protein SubtypeConcanavalin A (Con A)
Protein TagNone
Purity or Quality GradeImaging Quality
Quantity10 mg
Research CategoryImaging
Shipping ConditionRoom Temperature
SourceCanavlia ensiformis
ConjugateTexas Red
For Use With (Application)Immunocytochemistry, Immunohistochemistry, Immunofluorescence
FormLyophilized
RecombinantNative
SpeciesC. ensiformis
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (14)

Citations & References
Abstract
Exocytotic insertion of calcium channels constrains compensatory endocytosis to sites of exocytosis.
Authors:Smith RM, Baibakov B, Ikebuchi Y, White BH, Lambert NA, Kaczmarek LK, Vogel SS
Journal:J Cell Biol
PubMed ID:10684256
'Proteins inserted into the cell surface by exocytosis are thought to be retrieved by compensatory endocytosis, suggesting that retrieval requires granule proteins. In sea urchin eggs, calcium influx through P-type calcium channels is required for retrieval, and the large size of sea urchin secretory granules permits the direct observation of ... More
Identification of a novel domain in two mammalian inositol-polyphosphate 5-phosphatases that mediates membrane ruffle localization. The inositol 5-phosphatase skip localizes to the endoplasmic reticulum and translocates to membrane ruffles following epidermal growth factor stimulation.
Authors:Gurung R, Tan A, Ooms LM, McGrath MJ, Huysmans RD, Munday AD, Prescott M, Whisstock JC, Mitchell CA
Journal:J Biol Chem
PubMed ID:12536145
'SKIP (skeletal muscle and kidney enriched inositol phosphatase) is a recently identified phosphatidylinositol 3,4,5-trisphosphate- and phosphatidylinositol 4,5-bisphosphate-specific 5-phosphatase. In this study, we investigated the intracellular localization of SKIP. Indirect immunofluorescence and subcellular fractionation showed that, in serum-starved cells, both endogenous and recombinant SKIP colocalized with markers of the endoplasmic reticulum ... More
The N-terminus of retinol dehydrogenase type 1 signals cytosolic orientation in the microsomal membrane.
Authors:Wang J, Bongianni JK, Napoli JL
Journal:Biochemistry
PubMed ID:11601977
'We determined the orientation of the SDR (short-chain dehydrogenase/reductase) rat RoDH1 (retinol dehydrogenase type 1) in the endoplasmic reticulum to provide insight into its function in retinol metabolism, and to resolve whether retinoid-metabolizing SDRs differ from several other SDRs by requiring a C-terminal segment for the membrane orientation. In contrast ... More
Indirect immunofluorescence localization of ponticulin in motile cells.
Authors:Wuestehube LJ, Chia CP, Luna EJ
Journal:Cell Motil Cytoskeleton
PubMed ID:2673549
'Ponticulin is the major actin-binding integral glycoprotein in plasma membranes isolated from log-phase Dictyostelium discoideum amebae. As such, this protein appears to be an important link between the plasma membrane and actin filaments (Wuestehube and Luna: Journal of Cell Biology 105:1741-1751, 1987). In this study, indirect immunofluorescence microscopy was used ... More
Functional expression, quantification and cellular localization of the Hxt2 hexose transporter of Saccharomyces cerevisiae tagged with the green fluorescent protein.
Authors:Kruckeberg AL, Ye L, Berden JA, van Dam K
Journal:Biochem J
PubMed ID:10191260
'The Hxt2 glucose transport protein of Saccharomyces cerevisiae was genetically fused at its C-terminus with the green fluorescent protein (GFP). The Hxt2-GFP fusion protein is a functional hexose transporter: it restored growth on glucose to a strain bearing null mutations in the hexose transporter genes GAL2 and HXT1 to HXT7. ... More