Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TAP Tag Antibodies

          Invitrogen

          TAP Tag Polyclonal Antibody

          Advanced Verification
          85 Published Figures
          214 References
          View all (6) TAP Tag antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TAP Tag Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (85)
          • Advanced Verification (1)
          TAP Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TAP Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 90

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TAP Tag Antibody (CAB1001) in WB

          Western blot was performed using Anti-TAP Tag Polyclonal Antibody (Product # CAB1001) by loading whole cell extracts of untransfected and transiently transfected HEK-293E lysates: untransfected, 50 µg (Lane 1), empty vector control, 50 µg (Lane 2), H3-TAP, 50 µg (Lane 3), H3-TAP, 25 µg (Lane 4), H3-TAP, 12.5 µg (Lane 5), H3-TAP, 6.25 µg (Lane 6), p65-TAP, 50 µg (Lane 7) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # LC200... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Immunoprecipitation (IP)
          TAP Tag Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, ChIP Assay (WB, ChIP)
          TAP Tag Antibody in Western Blot, ChIP Assay (WB, ChIP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation, Dot Blot (WB, IP, DB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, ChIP Assay (WB, ChIP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation, ChIP Assay (WB, IP, ChIP)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Western Blot, Immunoprecipitation (WB, IP)
          TAP Tag Antibody in Western Blot (WB)
          TAP Tag Antibody in Immunoprecipitation (IP)
          TAP Tag Antibody in Immunoprecipitation (IP)
          TAP Tag Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          CAB1001

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1000
          View 151 publications 151 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 27 publications 27 publications

          ChIP assay (ChIP)

          Assay-dependent
          View 24 publications 24 publications

          Dot blot (DB)

          -
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Tag

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          BBS, pH 8.4

          Contains

          no preservative

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10709700

          Product Specific Information

          CAB1001 detects TAP.

          CAB1001 has been successfully used in Western Blot, IP, and ChIP applications

          The CAB1001 immunogen is a KLH conjugated peptide representing the C-terminus of the TAP construct after TEV cleavage.

          TAP adds about 20 kDa to the size of the protein.

          Target Information

          Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. Tandem affinity purification (TAP) is an affinity purification method for isolation of TAP-tagged proteins together with associated proteins. The protocol involves the fusion of the "TAP tag" (typically a calmodulin binding peptide (CBP), a tobacco etch virus protease (TEV protease) cleavage site and Protein A) to the protein of interest. The TAP technique is useful in analyzing in vivo interactions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Tobacco Etch Virus

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-xq5gc:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0