Barnstead™ TII Type 2 Water System
Actual product may vary
Thermo Scientific™

Barnstead™ TII Type 2 Water System

This item is no longer available and has been replaced by the next generation of Barnstead Water Purification Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
D14041Descontinued; replacement product is 50132132
D14031Discontinued; replacement product is 50132131
Catalog number D14041
Price (USD)
-
Description:
Descontinued; replacement product is 50132132

This item is no longer available and has been replaced by the next generation of Barnstead Water Purification Systems. The recommended replacement for this product is the Thermo Scientific™ Barnstead™ Pacific™ TII water purification system.

Produce Type 2 pure water that exceeds ASTM and CLSI-CLRW standards with the smart and flexible Thermo Scientific™ Barnstead™ TII Type 2 Water System. This system is ideal for all applications requiring pure water such as the preparation of microbiological media and buffers, glassware washing and as feed water for Type 1 ultrapure water systems.

Features:

  • Green/yellow/red indicator lights simplify operation with detailed status on alert information.
  • System is available in 12L and 24L/hr. production rates with 30L or 60L storage tanks for a variety of application needs.
  • System monitors water at all points including feedwater, RO water, final product water.
  • System measures resistivity and temperature.
  • Removable display can be mounted up to 10' (3m) from unit for added flexibility.
  • Space-saving design: unit can be wall-mounted or placed on benchtop UV lamp status can be monitored on display to ensure bacterial-free, low-organic water.
  • Cartridge disconnects easily for simple, fast replacement.
  • Fluorinated PE reservoir eliminates leaching of organic materials and protects against contamination.
  • Direct dispensing offers options of volumetric (0.25 to 60L) or timed dispense.

Smart, Easy to Use, Low-Maintenance System:

  • Multi-step purification combines pretreatment, reverse osmosis, deionization, UV oxidation and filtration in one.
  • System easily dispenses from unit or reservoir.
  • Exclusive flow pattern and unique water storage and recirculation system continuously maintain stored water.
  • Smart controls monitor critical system data and automate operation, ensuring maximum water quality.
  • Sophisticated multifunctional display provides real-time information on water quality, system performance and diagnostics.
  • Intuitive multifunction main menu has push-button controls to monitor critical system data.
  • System allows easy access to alarm information, setup data and real-time operational readings such as water purity, percent RO rejection, cartridge status and reservoir levels.

Ordering Information:
Optional accessories include remote dispensers, printer, cutoff float, interlock cable, storage reservoirs, bottles and replacement consumables. These products are available separately. For details, contact your sales representative.

Includes:

  • Wall bracket
  • RO membrane(s)
  • UV lamp

Requires:

  • Start-Up Kit (D502138)
  • Storage reservoir

Recommended for:

  • Media prep
  • Buffer preps
  • Glassware washing
  • Equipment feed
  • Type 1 Water system feed
Specifications
Bacterial retention99%
Certifications/ComplianceCE, CSA
Conductivity0.067 to 0.1 µS/cm
Depth (English)18.5 in.
Depth (Metric)47 cm
DescriptionDescontinued; replacement product is 50132132
Electrical Requirements100/240 VAC 50/60 Hz
Feedwater Pressure30 to 100 psig (2 to 6.9 bar)
Feedwater Temperature (Metric)4°C to 40°C
Frequency50/60 Hz
Height (English)19.5 in.
Height (Metric)49.5 cm
Particles >0.2µM/mL<1 μM/mL
Product TypeWater System
Purified Water Flowrate Field1 L/min.
RO Flow Rate12 L/hr.
Voltage100/240 VAC
Width (English)13.5 in.
Width (Metric)34.3 cm
Bacterial Content<1 CFU/mL
Feed Water MonitoringNo
Feedwater SourceTap
Product Water TypeType 2
Remote DispenserOptional
Resistivity15 Mohm.cm at 25°C
TOC Monitor with UV Intensity MonitoringNo
Total Organic Carbon1 to 5 ppb
Unit SizeEach