BODIPY&trade; FL (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-<i>s</i>-Indacene-3-Propionic Acid)
Invitrogen™

BODIPY™ FL (4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Propionic Acid)

BODIPY™ FL (4,4-difluoro-5,7-dimetil-4-bora-3a,4a-diaza-s-indaceno-3-ácido propiónico)
Have Questions?
Número de catálogoCantidad
D2183
también denominado D-2183
5 mg
Número de catálogo D2183
también denominado D-2183
Precio (MXN)
-
Cantidad:
5 mg

The amine-reactive BODIPY™ FL propionic acid can be used to create green-fluorescent bioconjugates following carbodiimide-activation of the carboxylic acid on the dye and reaction with amines. The electronically neutral BODIPY™ FL dye has the most fluorescein-like spectra of the green-fluorescent BODIPY™ dyes.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Reactividad químicaAmina
Etiqueta o tinteBODIPY™ FL
Tipo de productoÁcido propiónico FL
Cantidad5 mg
Fracción reactivaÁcido carboxílico
Condiciones de envíoTemperatura ambiente
Tipo de etiquetaColorantes BODIPY
Línea de productosBODIPY
Unit SizeEach
Contenido y almacenamiento
Almacenar en el congelador (de – 5 a – 30 °C) y proteger de la luz.

Citations & References (16)

Citations & References
Abstract
Thousandfold signal increase using field-amplified sample stacking for on-chip electrophoresis.
Authors:Jung B, Bharadwaj R, Santiago JG
Journal:Electrophoresis
PubMed ID:14595694
'Field-amplified sample stacking (FASS) leverages conductivity gradients between a volume of injected sample and the background buffer to increase sample concentration. A major challenge in applying FASS to on-chip assays is the initial setup of high-conductivity gradient boundaries in the region of the injected sample volume. We have designed, fabricated, ... More
Inter- and intramolecular fluorescence quenching of organic dyes by tryptophan.
Authors:Marmé N, Knemeyer JP, Sauer M, Wolfrum J
Journal:Bioconjug Chem
PubMed ID:14624626
'Steady-state and time-resolved fluorescence measurements were performed to elucidate the fluorescence quenching of oxazine, rhodamine, carbocyanine, and bora-diaza-indacene dyes by amino acids. Among the natural amino acids, tryptophan exhibits the most pronounced quenching efficiency. Especially, the red-absorbing dyes ATTO 655, ATTO 680, and the oxazine derivative MR 121 are strongly ... More
Toxicity of organic fluorophores used in molecular imaging: literature review.
Authors:Alford R, Simpson HM, Duberman J, Hill GC, Ogawa M, Regino C, Kobayashi H, Choyke PL,
Journal:Mol Imaging
PubMed ID:20003892
'Fluorophores are potentially useful for in vivo cancer diagnosis. Using relatively inexpensive and portable equipment, optical imaging with fluorophores permits real-time detection of cancer. However, fluorophores can be toxic and must be investigated before they can be administered safely to patients. A review of published literature on the toxicity of ... More
Interaction of fluorescence labeled single-stranded DNA with hexameric DNA-helicase RepA: a photon and fluorescence correlation spectroscopy study.
Authors:Xu H, Frank J, Trier U, Hammer S, Schröder W, Behlke J, Schäfer-Korting M, Holzwarth JF, Saenger W
Journal:Biochemistry
PubMed ID:11401568
'Fluorescence correlation spectroscopy (FCS) was used to characterize the interaction of fluorescence labeled single-stranded DNA (ssDNA) with hexameric RepA DNA-helicase (hRepA) encoded by plasmid RSF1010. The apparent dissociation constants, Kd(app), for the equilibrium binding of 12mer, 30mer, and 45mer ssDNA 5''-labeled with BFL to hRepA dimer in the presence of ... More
Activation and inhibition of adenylyl cyclase isoforms by forskolin analogs.
Authors:Pinto C, Papa D, Hübner M, Mou TC, Lushington GH, Seifert R,
Journal:J Pharmacol Exp Ther
PubMed ID:18184830
'Adenylyl cyclase (AC) isoforms 1 to 9 are differentially expressed in tissues and constitute an interesting drug target. ACs 1 to 8 are activated by the diterpene, forskolin (FS). It is unfortunate that there is a paucity of AC isoform-selective activators. To develop such compounds, an understanding of the structure/activity ... More