BODIPY™ 493/503 NHS Ester (Succinimidyl Ester) - FAQs
BODIPY™ 493/503 NHS Ester (Succinimidyl Ester) - FAQs
View additional product information for BODIPY™ 493/503 NHS Ester (Succinimidyl Ester) - FAQs (D2191)
1 product FAQs found
What is the difference between BODIPY 493/503 (4,4-Difluoro-1,3,5,7,8-Pentamethyl-4-Bora-3a,4a-Diaza-s-Indacene) (Cat. No. D3922) and BODIPY 493/503 NHS Ester (Succinimidyl Ester) (Cat. No. D2191)?
BODIPY 493/503 NHS Ester (Succinimidyl Ester) (Cat. No. D2191) is the amine‐reactive derivative of BODIPY 493/503 (4,4‐Difluoro‐1,3,5,7,8‐Pentamethyl‐4‐Bora‐3a,4a‐Diaza‐s‐Indacene) (Cat. No. D3922). The reactive group allows for D2191 to be used as a labeling reagent to label proteins or other molecules to covalently attach the dye to accessible primary amines. If using this as a live cell reagent, the reactive group may allow for better retention as it will covalently attach to any primary amines on any cellular material it may come into contact with.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.