BODIPY™ FL-X NHS Ester (Succinimidyl Ester)
Invitrogen™

BODIPY™ FL-X NHS Ester (Succinimidyl Ester)

BODIPY™ FL-X dye is bright, green fluorescent dye with similar excitation and emission to fluorescein (FITC) or Alexa Fluor™ 488Read more
Have Questions?
Catalog NumberQuantity
D6102
also known as D-6102
5 mg
Catalog number D6102
also known as D-6102
Price (USD)
562.00
Each
Quantity:
5 mg
Request bulk or custom format
Price (USD)
562.00
Each
BODIPY™ FL-X dye is bright, green fluorescent dye with similar excitation and emission to fluorescein (FITC) or Alexa Fluor™ 488 dye. It has a high extinction coefficient and fluorescence quantum yield and is relatively insensitive to solvent polarity and pH change. In contrast to the highly water soluble fluorophores Alexa Fluor™ 488 dye and fluorescein (FITC), BODIPY™ dyes have unique hydrophobic properties ideal for staining lipids, membranes, and other lipophilic compounds. BODIPY™ FL-X dye has a relatively long excited-state lifetime (typically 5 nanoseconds or longer), which is useful for fluorescence polarization-based assays and a large two-photon cross-section for multiphoton excitation. In addition to reactive dye formulations, we offer BODIPY™ FL-X dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of BODIPY™ FL-X is the most popular tool for conjugating the dye to a protein or antibody. NHS esters can be used to label the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting BODIPY™ FL-X conjugates exhibit bright fluorescence, narrow emission bandwidths, and relatively long excited-state lifetimes, which can be useful for fluorescence polarization assays and two-photon excitation (TPE) microscopy.

This reactive dye contains a seven-atom aminohexanoyl ('X') spacer between the fluorophore and the NHS ester group. This spacer helps to separate the fluorophore from its point of attachment, potentially reducing the interaction of the fluorophore with the biomolecule to which it is conjugated.

Detailed information about this BODIPY™ FL-X NHS ester:

Fluorophore label: BODIPY™ FL-X dye
Reactive group: NHS ester (succinimidyl ester)
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 504/510 nm
Extinction coefficient: 85,000 cm-1M-1
Molecular weight: 502.32

Typical Conjugation Reaction
Amine-reactive reagents can be conjugated with virtually any protein or peptide; the provided protocol is optimized for IgG antibodies. The reaction can be scaled for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The BODIPY™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO), and the reaction is carried out in 0.1-0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free BODIPY™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration medium with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission510 nm
Excitation504 nm
Label or DyeBODIPY™ FL
Product TypeNHS Ester (Succinimidyl Ester)
Quantity5 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeBODIPY Dyes
Product LineBODIPY
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Citations & References (5)

Citations & References
Abstract
Fluorescent labeling of cell-free synthesized proteins by incorporation of fluorophore-conjugated nonnatural amino acids.
Authors:Kang SH, Jun SY, Kim DM
Journal:Anal Biochem
PubMed ID:17113028
'Although fluorescent dyes, such as fluorescein derivatives, have bulky and complex structures, nonnatural amino acids carrying these fluorescein derivatives are acceptable by the Escherichia coli ribosome and are useful for the cotranslational fluorescent labeling of cell-free synthesized proteins. Surprisingly, the incorporation efficiency of nonnatural amino acids carrying fluorescein derivatives into ... More
Controlling the intracellular localization of fluorescent polyamide analogues in cultured cells.
Authors:Crowley KS, Phillion DP, Woodard SS, Schweitzer BA, Singh M, Shabany H, Burnette B, Hippenmeyer P, Heitmeier M, Bashkin JK
Journal:Bioorg Med Chem Lett
PubMed ID:12699756
The intracellular distribution of fluorescent-labeled polyamides was examined in live cells. We showed that BODIPY-labeled polyamides accumulate in acidic vesicles, mainly lysosomes, in the cytoplasm of HCT116 colon cancer cells and human rheumatoid synovial fibroblasts (RSF). Verapamil blocked vesicular accumulation and led to nuclear accumulation of the BODIPY-labeled polyamide in ... More
Influence of fluorophore and linker composition on the pharmacology of fluorescent adenosine A1 receptor ligands.
Authors:Baker JG, Middleton R, Adams L, May LT, Briddon SJ, Kellam B, Hill SJ,
Journal:Br J Pharmacol
PubMed ID:20105183
BACKGROUND AND PURPOSE: The introduction of fluorescence-based techniques, and in particular the development of fluorescent ligands, has allowed the study of G protein-coupled receptor pharmacology at the single cell and single molecule level. This study evaluated how the physicochemical nature of the linker and the fluorophore affected the pharmacological properties ... More
A novel subfractionation approach for mitochondrial proteins: a three-dimensional mitochondrial proteome map.
Authors:Hanson BJ, Schulenberg B, Patton WF, Capaldi RA
Journal:Electrophoresis
PubMed ID:11332763
As mitochondria play critical roles in both cell life and cell death, there is great interest in obtaining a human mitochondrial proteome map. Such a map could potentially be useful in diagnosing diseases, identifying targets for drug therapy, and in screening for unwanted drug side effects. In this paper, we ... More
Binding and internalization of lipopolysaccharide by Cla-1, a human orthologue of rodent scavenger receptor B1.
Authors:Vishnyakova TG, Bocharov AV, Baranova IN, Chen Z, Remaley AT, Csako G, Eggerman TL, Patterson AP
Journal:J Biol Chem
PubMed ID:12651854
Scavenger receptor, class B, type I (SR-BI) mediates selective uptake of high density lipoprotein (HDL) cholesteryl ester. SR-BI recognizes HDL, low density lipoprotein (LDL), exchangeable apolipoproteins, and protein-free lipid vesicles containing negatively charged phospholipids. Lipopolysaccharides (LPS) are highly glycosylated anionic phospholipids contributing to septic shock. Despite significant structural similarities between ... More