Nalgene™ Spherical-Bottom Polycarbonate Centrifuge Bottle Adapter
Thermo Scientific™
Nalgene™ Spherical-Bottom Polycarbonate Centrifuge Bottle Adapter
Use this spherical-bottom centrifuge adaptor, molded of rigid, robust white polycarbonate for assured durability.
Have Questions?
Catalog NumberProduct Type
DS3124-0010Centrifuge Adapter
Catalog number DS3124-0010
Price (USD)
102.85
Online exclusive
127.00
Save 24.15
Case of 4
Product Type:
Centrifuge Adapter
Price (USD)
102.85
Online exclusive
127.00
Save 24.15
Case of 4
Use the Thermo Scientific™ Nalgene™ Spherical-Bottom Polycarbonate Centrifuge Bottle Adapter, specifically designed to mate with the Nalgene Spherical-Bottom Polycarbonate Centrifuge Bottle (3123-0250) for use in standard centrifuge rotors. It is molded of rigid and robust white polycarbonate for assured durability.
For use with Nalgene 3123-0250 Round bottom polycarbonate centrifuge bottles.
Adapts the Nalgene round bottom bottle to fit in standard centrifuge rotors.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Specifications
Closure MaterialPolycarbonate
For Use With (Equipment)Standard Centrifuge Rotors, Spherical Bottom Centrifuge Bottle
Height (English)5.41 in.
Height (Metric)137.5 mm
No. per Case4
No. per Pack4
Product TypeCentrifuge Adapter
ShapeRound
ColorWhite
MaterialPolycarbonate
Unit SizeCase of 4