Can I use the Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method) (Cat. No. EEA026) with cell samples?
We do not recommend using the Total Cholesterol (TC) Colorimetric Assay Kit with cell samples because most untreated cell samples have a low TC that does not meet the sensitivity of the kit.
Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .
Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .