Search
Search

| Catalog Number | Quantity | Concentration | Product Type |
|---|---|---|---|
| EF0221T1 | 10 U, 10 μL | 1 U/μL | Pyrophosphatase, Inorganic |
| EN052T1 | 5000 U, 100 μL | 50 U/μL | DNase I, RNase-free |
| EO038T1 | 10,000 U, 250 μL | 40 U/μL | RNase Inhibitor |
| EF022T1 | 10 U, 100 μL | 0.1 U/μL | Pyrophosphatase, Inorganic |
The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.
Features
The exceptional quality of TheraPure products is supported by: