Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • CD85j (ILT2) ELISA Kits

          Invitrogen

          Human ILT-2/LIR-1/CD85j ELISA Kit

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human ILT-2/LIR-1/CD85j ELISA Kit
          Group 53 Created with Sketch.
          Human ILT-2/LIR-1/CD85j ELISA Kit
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human ILT-2/LIR-1/CD85j ELISA Kit

          ELISA Standard curve of ILT-2/LIR-1/CD85j using Human ILT-2/LIR-1/CD85j ELISA Kit (Product # EH277RB). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human ILT-2/LIR-1/CD85j ELISA Kit
          Human ILT-2/LIR-1/CD85j ELISA Kit (EH277RB)
          videoThumbnail
          ►

          Product Specifications

          Analytical sensitivity

          22 pg/mL

          Assay range

          22-5,000 pg/mL

          Sample type/volume

          Supernatant
          50 µL
          Serum
          50 µL
          Plasma
          50 µL

          Hands-on time

          1 hr 20 min

          Time-to-result

          4 hr 45 min

          Homogenous (no wash)

          No

          Interassay CV

          <12%

          Intraassay CV

          <10%

          Instrument

          Absorbance Microplate Reader

          Product size

          96 Tests

          Contents

          Pre-coated 96 well plate
          Standard
          Assay Diluent concentrate
          Biotinylated Detection Antibody
          SAV-HRP
          Wash Buffer
          Chromogen
          Stop Solution
          Adhesive Plate Covers

          Shipping conditions

          Wet ice

          Storage

          Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C

          Protein name

          CD85j (ILT2)

          Species (tested)

          Human

          Detector antibody conjugate

          Biotin

          Label or dye

          HRP

          About This Kit

          Human ILT-2/LIR-1/CD85j quantitates human ILT-2/LIR-1/CD85j in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ILT-2/LIR-1/CD85j.

          Target information

          CD85j, also known as a member of the leukocyte immunoglobulin-like receptor (LIR) family, is a single transmembrane glycoprotein with a long cytoplasmic domain containing four immunoreceptor tyrosine-based inhibitory motifs (ITIMs). These motifs signal through interactions with SHP-1, transducing negative signals that inhibit immune response stimulation. CD85j is expressed on myeloid cells such as monocytes and dendritic cells, as well as on subsets of lymphoid cells including NK, T, and B cells. The expression of CD85j is correlated with leukemias such as acute lymphoblastic leukemia (ALL) and chronic lymphocytic leukemia (CLL). On CD8+ cells, CD85j expression is associated with effector cell function and plays a significant role in viral infections, including HIV, Epstein-Barr virus, and cytomegalovirus (CMV). CD85j binds to MHC class I molecules, including HLA-G, A, F, B27, E, and F, on antigen-presenting cells. The receptor helps control inflammatory responses and cytotoxicity, focusing the immune response and limiting autoreactivity. CD85j belongs to the subfamily B class of LIR receptors, which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and multiple transcript variants encoding different isoforms have been identified for this gene. Its gene is located in a cluster at chromosomal region 19q13.4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Gene aliases : CD85J, ILT-2, ILT2, LIR-1, LIR1, MIR-7, MIR7, PIR-B, PIRB

          Gene ID : (Human) 10859

          Gene symbol : LILRB1

          Protein Aliases : CD85 antigen-like family member J, Ig-like transcript 2, ILT2a, allelic form of ILT2, ILT2b, alternatively spliced form of ILT2, ILT2c, immunoglobulin heavy chain variable region, leucocyte Ig-like receptor B1, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1, LIR-1, monocyte/macrophage immunoglobulin-like receptor 7, myeloid inhibitory receptor 7, unnamed protein product

          UniProt ID (Human) Q8NHL6

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0