Informational:Regulatory requirements vary by country; products may not be available in your geographic area.
Thermo Scientific™

VetMAX™ EHDV Kit

Catalog number: EHDV50
Thermo Scientific™

VetMAX™ EHDV Kit

Catalog number: EHDV50
Catalog Number
EHDV50
Unit Size
50 tests
Price (USD)
Catalog NumberUnit SizePrice (USD)
EHDV5050 testsContact Us ›
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Note: The name of this product has changed. It was previously called LSI VetMAX Epizootic Hemorrhagic Disease Virus.

Same as TaqVet Epizootic Hemorrhagic Disease Virus Real-Time PCR Kit

Real-time PCR enables sensitive and specific detection of pathogen nucleic acid in animal samples, allowing for reliable and rapid screening and detection of infected animals.

The LSI VetMAX™ kits include:
• 1 ready-to-use mix that includes a set of oligonucleotides for detection of the target gene OR these separate components that will need to be mixed together: a set of oligonucleotides for the detection of the target gene + a TaqMan™ real-time PCR master mix + an RT enzyme (only for RNA kits).
• 1 external positive control of the same nature as the target (inactivated bacteria or virus). Use as a quality control for PCR and/or extraction yield.
• A quality control certificate.

For veterinary use only.

Regulatory requirements vary by country; products may not be available in your geographic area.

Biological hazard safety

WARNING! BIOHAZARD. Biological samples such as tissues, body fluids, infectious agents, and blood of humans and other animals have the potential to transmit infectious diseases. All work should be conducted in properly equipped facilities using the appropriate safety equipment (for example, physical containment devices). Safety equipment also may include items for personal protection, such as gloves, coats, gowns, shoe covers, boots, respirators, face shields, safety glasses, or goggles. Individuals should be trained according to applicable regulatory and company/institution requirements before working with potentially biohazardous materials. Follow all applicable local, state/provincial, and/or national regulations. The following references provide general guidelines when handling biological samples in laboratory environment.

U.S. Department of Health and Human Services, Biosafety in Microbiological and Biomedical Laboratories (BMBL), 5th Edition, HHS Publication No. (CDC) 21-1112, Revised December 2009

World Health Organization, Laboratory Biosafety Manual, 3rd Edition, WHO/CDS/CSR/LYO/2004.11

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
Low
Product Line
VetMAX™
Product Type
PCR Reagents and Detection Kit for Animal Health
Species
Cattle/Bovine, Goat/Caprine, Sheep, Sheep/Ovine, Wild Ruminant
Target Organism Class
Epizootic Hemorrhagic Disease Virus (EHDV)
For Use With (Application)
Real Time PCR (qPCR)
No. of Reactions
1 Well Reaction
Sample Type
Abortive Animal, Whole Blood (EDTA tube), spleen, Spleen

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text