Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • cGAMP ELISA Kits

          Invitrogen

          2'3'- Cyclic GAMP ELISA Kit

          5 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          2'3'- Cyclic GAMP ELISA Kit (EIAGAMP)
          Group 53 Created with Sketch.
          2'3'- Cyclic GAMP ELISA Kit (EIAGAMP)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          2'3'- Cyclic GAMP ELISA Kit

          Competitive ELISA {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          2'3'- Cyclic GAMP ELISA Kit (EIAGAMP)
          videoThumbnail
          ►

          Product Specifications

          Analytical sensitivity

          0.048 pmol/mL

          Assay range

          0.08-20 pmol/mL

          Sample type/volume

          Cell lysate
          50 µL
          Tissue homogenate
          50 µL
          Supernatant
          50 µL
          Plasma
          10 µL

          Hands-on time

          45 min

          Time-to-result

          2 hr 30 min

          Homogenous (no wash)

          No

          Interassay CV

          8%

          Intraassay CV

          6%

          Instrument

          Absorbance Microplate Reader

          Product size

          96 Tests

          Contents

          Anti-IgG coated 96 well plate
          Standard
          Assay Buffer Concentrate
          Capture Antibody
          HRP-conjugated Antigen
          Wash Buffer Concentrate
          TMB Substrate
          Stop Solution
          Plate Sealer

          Shipping conditions

          Dry ice

          Storage

          2-8°C

          Protein name

          cGAMP

          Species (tested)

          Human

          Detector antibody conjugate

          HRP

          Label or dye

          Unconjugated

          About This Kit

          The 2',3'-Cyclic GAMP Competitive ELISA Kit quantitates 2',3'-cyclic GAMP in lysed cells and tissue, tissue culture media, and EDTA plasma.|Principle of the method: The 2',3'-Cyclic GAMP Competitive ELISA research-use-only kit is designed to quantitatively measure 2',3'-cyclic GAMP independent of species. A 2',3'-cyclic GAMP standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit 2',3'-cyclic GAMP antibodies and 2',3'-cyclic GAMP-peroxidase conjugate is added to the wells. The 2',3'-cyclic GAMP-peroxidase conjugate and any 2',3'-cyclic GAMP in the sample will compete to bind to the anti-rabbit 2',3'-cyclic GAMP antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound 2',3'-cyclic GAMP-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of 2',3'-cyclic GAMP present in the sample.|Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

          Target information

          Cyclic GMP-AMP (cGAMP) synthase (cGAS) is a nucleotidyltransferase located in the cytosol that acts as a cytosolic DNA sensor to detect foreign DNA from microbial pathogens as part of the innate immune response. Upon binding to cytosolic DNA, cGAS produces the cyclic dinucleotide second messenger cGAMP, which activates stimulator of interferon genes (STING), leading to activation of the type I interferon (IFN) pathway. In vitro, fibroblasts, macrophages, and dendritic cells isolated from cGAS knockout (cGAS-/-) mice do not produce type I IFNs following DNA transfection or DNA virus infection. Similarly, cells containing a frame-shift mutation in the cGAS locus fail to mount an immune response to HIV and other retroviruses. In vivo, cGAS-/- mice infected with herpes simplex virus 1 (HSV-1) have lower levels of IFN-α and IFN-β, shorter survival times, and higher post-mortem levels of HSV-1 in the brain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases : 2'3'-Cyclic GMP-AMP, cGAMP, Guanosine-Adenosine 2', 3'-cyclic monophosphate

          Recommended Products

          IL-6 Human Matched Antibody Pair

          Because you are interested in IL-6 Human ELISA Kit.

          SIZE

          10 x 96 tests

          PRICE

          USD 400.00

          Cat # CHC1263

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0