Search
View additional product information for Glucose Colorimetric Detection Kit - FAQs (EIAGLUC)
1 product FAQs found
The Glucose Colorimetric Detection Kit (Cat. No. EIAGLUC) is designed to measure glucose in various sample types, including serum, plasma, urine, buffered solutions, and tissue culture medium. While the assay was characterized with human samples, the kit is described as species-independent, meaning it should be suitable for use with samples from other species including mouse samples.
However, it is always recommended to validate the assay with your specific sample type to help ensure accurate and reliable results.