RNase T1 (1000 U/μL)
Thermo Scientific™

RNase T1 (1000 U/μL)

Thermo Scientific RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues. It cleaves the phosphodiester bondRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EN0541100,000 units
EN0542500,000 units
Catalog number EN0541
Price (USD)
86.65
Online exclusive
103.00
Save 16.35
Each
Quantity:
100,000 units
Request bulk or custom format
Price (USD)
86.65
Online exclusive
103.00
Save 16.35
Each
Thermo Scientific RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues. It cleaves the phosphodiester bond between the 3'-guanylic residue and the 5'-OH residue of adjacent nucleotides with the formation of corresponding intermediate 2', 3'-cyclic phosphate. The reaction products are 3'-GMP and oligonucleotides with a terminal 3'-GMP.

RNase T1 does not require metal ions for activity.

Applications

• Removal of RNA from DNA preparations
• RNA sequencing
• Ribonuclease protection assays. Used in conjunction with RNase A
• Removal of RNA from recombinant protein preparations
• Determination of the level of RNA transcripts synthesized in vitro from DNA templates containing a 'G-less cassette'
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeRNase T1
Quantity100,000 units
Concentration1000 U/μL
EnzymeRNase
Unit SizeEach

Frequently asked questions (FAQs)

How much Thermo Scientific RNase T1 should I use for complete digestion of RNA?

We would recommend 0.2 µL RNase T1 per µg of RNA in 50 mM Tris-HCl (pH 7.5) and 1 mM EDTA. Incubate at 37 degrees C for 30 min.