Lambda Exonuclease (10 U/μL)
Thermo Scientific™

Lambda Exonuclease (10 U/μL)

Thermo Scientific Lambda Exonuclease is a highly processive 5'→3' exodeoxyribonuclease. It selectively digests the 5'-phosphorylated strand of double-stranded DNA. TheRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EN05625,000 units
EN05611,000 units
Catalog number EN0562
Price (USD)
401.65
Online exclusive
437.00
Save 35.35
Each
Quantity:
5,000 units
Request bulk or custom format
Price (USD)
401.65
Online exclusive
437.00
Save 35.35
Each
Thermo Scientific Lambda Exonuclease is a highly processive 5'→3' exodeoxyribonuclease. It selectively digests the 5'-phosphorylated strand of double-stranded DNA. The enzyme exhibits low activity on single-stranded DNA and non-phosphorylated DNA, and has no activity at nicks and limited activity at gaps in DNA.

Highlights

• Active in Thermo Scientific PCR buffers

Applications
• Generating single-stranded PCR products for use in: • DNA sequencing
• Analysis of DNA single-strand conformation polymorphism (SSCP)
• Rolling circle amplification

• Producing single-stranded DNA from double-stranded DNA fragments
• Cloning of PCR products

Note

Use of this enzyme in certain applications may be covered by patents and may require a license.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeExonuclease
Quantity5,000 units
Concentration10 U/μL
EnzymeExonuclease
Unit SizeEach