TheraPure™ Modifying Enzymes
Thermo Scientific™

TheraPure™ Modifying Enzymes

Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentrationProduct Type
EO038T110,000 U, 250 μL40 U/μLRNase Inhibitor
EN052T15000 U, 100 μL50 U/μLDNase I, RNase-free
EF0221T110 U, 10 μL1 U/μLPyrophosphatase, Inorganic
EF022T110 U, 100 μL0.1 U/μLPyrophosphatase, Inorganic
Catalog number EO038T1
Price (USD)
471.00
Each
Quantity:
10,000 U, 250 μL
Concentration:
40 U/μL
Product Type:
RNase Inhibitor
Request bulk or custom format
Price (USD)
471.00
Each
Thermo Scientific TheraPure in vitro transcription (IVT) enzymes are designed for advanced research and early development of mRNA therapeutics and vaccines. These products offer full workflow solutions from template preparation to RNA synthesis and are recommended for process research and optimization, proof of concept, and preclinical studies.

The high purity of TheraPure IVT enzymes is ensured by precise quality testing that includes confirmation that that the enzymes are nuclease- and endotoxin-free. In addition, the manufacturing process of TheraPure products follows ISO9001 and ISO13485 standards.

Features

  • Convenient stand-alone enzymes for easy workflow optimization
  • Animal-origin-free (AOF) raw materials used in manufacturing
  • No risk of prion or animal virus contamination
  • Reproducible and consistent results with validated quality testing
  • Regulatory compliant—β-lactam-free manufacturing technology
  • Suitable for translational research
  • Facilitates transition to GMP during mRNA vaccine development process
  • Scalable and available as OEM product

The exceptional quality of TheraPure products is supported by:

  • Certificates of analysis
  • Certificates of origin
  • TSE/BSE statement
For Research Use Only.
Specifications
For Use With (Application)In vitro Transcription
Inactivation Temperature75°C
InhibitorsSDS, urea, and all oxidizing reagents (p-chloromercuribenzoate, dissolved oxygen, ions) in their higher oxidation states
Product TypeRNase Inhibitor
Quantity10,000 U, 250 μL
Concentration40 U/μL
EnzymeRNase Inhibitor
FormLiquid
Product LineTheraPure
Research CategoryDrug Development
Sensitive to Heat InactivationYes
Unit SizeEach