Klenow Fragment (10 U/μL)
Thermo Scientific™

Klenow Fragment (10 U/μL)

Thermo Scientific Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'→3' polymerase activity and 3'→5' exonucleaseRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentration
EP00521,500 units10 U/μL
EP0051300 units10 U/μL
Catalog number EP0052
Price (USD)
545.65
Online exclusive
620.00
Save 74.35
Each
Quantity:
1,500 units
Concentration:
10 U/μL
Request bulk or custom format
Price (USD)
545.65
Online exclusive
620.00
Save 74.35
Each
Thermo Scientific Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'→3' polymerase activity and 3'→5' exonuclease (proofreading) activity, but lacks 5'→3' exonuclease activity of DNA polymerase I.

Highlights

• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
• Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers

Applications
• DNA blunting by fill-in 5'-overhangs
• Random-primed DNA labeling
• Labeling by fill-in 5'-overhangs of dsDNA
• DNA sequencing by the Sanger method
• Site-specific mutagenesis of DNA with synthetic oligonucleotides
• Second strand synthesis of cDNA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
PolymeraseDNA Polymerase I
Product TypeKlenow Fragment
Quantity1,500 units
Concentration10 U/μL
Unit SizeEach

Frequently asked questions (FAQs)

What are the differences between Thermo Scientific DNA Polymerase I, Klenow Fragment, and Klenow Fragment (exo-)?

Thermo Scientific DNA Polymerase I possesses 5′?3′ DNA synthesis activity, 3′?5′ exonuclease (proofreading) activity, and 5′?3′ exonuclease activity. Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5′?3′ polymerase activity and 3′?5′ exonuclease (proofreading) activity, but lacks 5′?3′ exonuclease activity. Klenow Fragment, exo-, is also the large fragment of DNA polymerase I. It exhibits 5′?3′ polymerase activity, but lacks the 3′?5′ and 5′?3′ exonuclease activities.

What conditions are optimal for fill-in reactions using Klenow (Large fragment of DNA Polymerase)?

Klenow can be used to "fill in" 5' overhangs of double-stranded DNA fragments using common restriction endonuclease buffers. We recommend REact 2 buffer (1X concentration is 50 mM Tris-HCl pH 8, 50 mM NaCl, 10 mM MgCl2) , but other buffers will also work.

Fill-in Reaction Conditions:

1. Dilute Large Fragment of DNA Polymerase I to 0.5 U/µL with Klenow Dilution Buffer.
2. To a 1.5-mL microcentrifuge tube on ice, add: 10X REact 2 Buffer - 3 µL, 0.5 mM dATP - 1 µL, 0.5 mM dCTP - 1 µL, 0.5 mM dGTP - 1 µL, 0.5 mM dTTP - 1 µL, DNA 0.5-1 µg, Large fragment of DNA Polymerase I - 1 µL, Autoclaved distilled water to 30 µL.
3. Mix gently and centrifuge briefly to bring the contents to the bottom of the tube.
4. Incubate at room temperature for 10-15 minutes or 20 minutes on ice.
5. Terminate fill-in reaction by phenol extraction.

To label the DNA fragment, use 1-2 µL of [alpha-32P]dNTP (400 Ci/mmol, 10 mCi/mL) (24-48 pmoles) instead of the corresponding cold dNTP.

Note: Thermo Fisher Scientific also offers an Exo minus Klenow, which is provided with its own buffers.