Fura-2, Pentapotassium Salt, cell impermeant
Invitrogen™

Fura-2, Pentapotassium Salt, cell impermeant

Fura-2, pentapotassium salt is an intracellular calcium indicator that is ratiometric and UV light-excitable.

Have Questions?
Catalog NumberQuantity
F12001 mg
Catalog number F1200
Price (USD)
247.65
Online exclusive
263.00
Save 15.35
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
247.65
Online exclusive
263.00
Save 15.35
Each

Fura-2, pentapotassium salt is an intracellular calcium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (Cat. No. I14402).

Fura-2, pentapotassium salt is an intracellular calcium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (Cat. No. I14402).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeFura-2
Emission Wavelength364 nm
Excitation Wavelength339 nm
Quantity1 mg
Shipping ConditionRoom Temperature
Emission Wavelength Range360 to 375 nm
Excitation Wavelength Range330 to 345 nm
For Use With (Application)Calcium Indicator
For Use With (Equipment)Fluorescence Microscope
Product TypeFura-2
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C) and protect from light.

Citations & References (751)

Citations & References
Abstract
Simultaneous measurement of intracellular pH and Ca2+ in insulin-secreting cells by spectral imaging microscopy.
Authors:Martínez-Zaguilán R,Gurulé MW,Lynch RM
Journal:The American journal of physiology
PubMed ID:8967445
Arabidopsis abi1-1 and abi2-1 phosphatase mutations reduce abscisic acid-induced cytoplasmic calcium rises in guard cells.
Authors:Allen GJ,Kuchitsu K,Chu SP,Murata Y,Schroeder JI
Journal:The Plant cell
PubMed ID:10488243
Elevations in cytoplasmic calcium ([Ca(2)+](cyt)) are an important component of early abscisic acid (ABA) signal transduction. To determine whether defined mutations in ABA signal transduction affect [Ca(2)+](cyt) signaling, the Ca(2)+-sensitive fluorescent dye fura 2 was loaded into the cytoplasm of Arabidopsis guard cells. Oscillations in [Ca(2)+](cyt) could be induced when ... More
Ca2+ transients in cardiac myocytes measured with high and low affinity Ca2+ indicators.
Authors:Berlin JR,Konishi M
Journal:Biophysical journal
PubMed ID:8274651
Intracellular calcium ion ([Ca2+]i) transients were measured in voltage-clamped rat cardiac myocytes with fura-2 or furaptra to quantitate rapid changes in [Ca2+]i. Patch electrode solutions contained the K+ salt of fura-2 (50 microM) or furaptra (300 microM). With identical experimental conditions, peak amplitude of stimulated [Ca2+]i transients in furaptra-loaded myocytes ... More
Evidence for role of cytosolic free calcium in hypoxia-induced proximal tubule injury.
Authors:Kribben A, Wieder ED, Wetzels JF, Yu L, Gengaro PE, Burke TJ, Schrier RW
Journal:J Clin Invest
PubMed ID:8182125
The role of cytosolic free Ca2+ ([Ca2+]i) in hypoxic injury was investigated in rat proximal tubules. [Ca2+]i was measured using fura-2 and cell injury was estimated with propidium iodide (PI) in individual tubules using video imaging fluorescence microscopy. [Ca2+]i increased from approximately 170 to approximately 390 nM during 5 min ... More
Hyposmotic activation hyperpolarizes outer hair cells of guinea pig cochlea.
Authors:Harada N, Ernst A, Zenner HP
Journal:Brain Res
PubMed ID:8348313
The electrophysiological responses of isolated guinea pig outer hair cells (OHCs) to hyposmotic activation were studied using the whole-cell patch-clamp technique. The cell swelling by hyposmotic activation hyperpolarized OHCs by 6.6 +/- 2.3 mV from the resting membrane potential of -58.5 +/- 5.9 mV (n = 48). This hyperpolarization was ... More