Can I fix my cells after loading with Fluo-4 AM dye for the detection of calcium?
No. Since Fluo-4 AM isn't covalently bound to any cellular components and fixation compromises the membrane, the dye would not be retained by the cell.