FluoroMyelin™ Red Fluorescent Myelin Stain - Solution in Water - FAQs
View additional product information for FluoroMyelin™ Red Fluorescent Myelin Stain - Solution in Water - FAQs (F34652)
4 product FAQs found
I used one of your FluoroMyelin stains and noticed it stains cells other than glial cells. Is there something wrong with the product?
FluoroMyelin is a lipid stain, any lipid can be stained by it but there is a higher lipid content in myelin that it will stain much more intensely than other membranes.
Can the FluoroMyelin stains be used on neurons in culture?
We have only tested them on mouse brain cryosections. They might work on neurons in culture, but would likely stain the membrane and there would be some endocytosis of the stains. Myelin sheaths may show enhanced intensity.
Can the FluoroMyelin myelin stains be used on paraffin sections?
We have only tested them on mouse brain cryosections.
What are the FluoroMyelin myelin stains?
They are proprietary lipid stains that stain myelin, but are not neuron specific.