Product Image
Thermo Scientific™

Phusion™ HF and GC Buffer Packs

5X Phusion Green buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberChemical Name or MaterialColor
F521LDetergent-free GC Buffer PackColorless
F518LHF Buffer PackColorless
F519LGC Buffer PackColorless
F520LDetergent-free FF Buffer PackColorless
F538LHF Buffer PackGreen
F539LGC Buffer PackGreen
Catalog number F521L
Price (USD)
44.65
Online exclusive
48.33
Save 3.68
Each
Chemical Name or Material:
Detergent-free GC Buffer Pack
Color:
Colorless
Request bulk or custom format
Price (USD)
44.65
Online exclusive
48.33
Save 3.68
Each
The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.

5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases:

  • 5X Phusion HF Buffer (F-518L, F-538L (Phusion Green), F-520L (Detergent-free))
  • 5X Phusion GC Buffer (F-519L, F-539L (Phusion Green), F-521L (Detergent-free))
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialDetergent-free GC Buffer Pack
ColorColorless
DescriptionDetergent-free Phusion™ GC Buffer Pack
Includes• 5X Phusion GC Reaction Buffer, Detergent-free (4 x 1.5 mL)
• 50 mM MgCl2 (4 x 1.5 mL)
• DMSO (0.5 mL)

Store at –20°C.
For Use With (Application)Standard PCR
Quantity4 x 1.5 mL
Unit SizeEach