Phusion™ High-Fidelity DNA Polymerases (2 U/μL)
Thermo Scientific™

Phusion™ High-Fidelity DNA Polymerases (2 U/μL)

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호색상반응 수
F530LColorless500 Reactions
F530SColorless100 Reactions
F-530XLColorless2000 Reactions
F534SGreen100 Reactions
F534LGreen500 Reactions
카탈로그 번호 F530L
제품 가격(KRW)
700,000
온라인 행사
Ends: 31-Dec-2026
777,000
할인액 77,000
Each
색상:
Colorless
반응 수:
500 Reactions
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
700,000
온라인 행사
Ends: 31-Dec-2026
777,000
할인액 77,000
Each
Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is an excellent choice for cloning and other applications requiring high fidelity. Phusion DNA polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerases.

Features of Phusion™ High-Fidelity DNA Polymerase include:

  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15–30 s/kb)
  • Robust performance, minimal optimization needed
  • High yields of PCR products with minimal enzyme amounts
  • Available in Green buffer format for direct loading of PCR products on gels (F-534S or F-534L)

Applications

  • High-fidelity PCR
  • Cloning
  • Template generation for sequencing
  • Amplification of difficult (GC-rich) templates
  • Long-range PCR (up to 20 kb)
  • Mutagenesis
  • High throughput PCR
  • Microarray

Note: Annealing rules for Phusion DNA polymerases are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use our convenient Tm calculator.

For Research Use Only. Not for use in diagnostic procedures.
사양
색상Colorless
Fidelity (Taq 대비)52X
핫 스타트No
반응 수500 Reactions
오버행Blunt
중합효소Phusion High-Fidelity DNA Polymerase
제품 유형High-Fidelity DNA Polymerase
수량500 units
반응 형식Standalone
배송 조건Dry Ice
크기(최종 제품)20 kb or less
농도2 U/μL
용도(애플리케이션)Standard PCR, High-fidelity PCR
GC-Rich PCR PerformanceHigh
반응 속도Fast
Unit SizeEach
구성 및 보관
• Phusion DNA Polymerase (2 U/μL)
• 5X Phusion HF & GC Buffers
• DMSO
• 50 mM MgCl2 solution

Phusion HF Buffer and Phusion GC Buffers each provide 1.5 mM MgCl2 in the final 1X concentration.
Store at -20°C.

자주 묻는 질문(FAQ)

What is enzyme concentration in Phusion Hot Start II High-Fidelity PCR Master Mix?

Phusion Hot Start II DNA polymerase concentration is optimized to give good results in most reactions. When the PCR reaction is set up according to the instructions, the final concentration of Phusion enzyme is 1 U in 50 µL reaction (0.4 U in 20 µL reaction).

Do Phusion DNA Polymerases add the non-template dependent 3'-A overhang?

Phusion DNA Polymerases generate blunt end products; therefore, blunt end cloning is recommended. If TA cloning is required, it can be performed by adding A overhangs to the blunt PCR product with e.g. Taq DNA Polymerase (Cat. No. EP0401). However, before adding the overhangs it is very important to remove all the Phusion DNA Polymerase by purifying the PCR product carefully, as the proofreading activity in Phusion DNA Polymerase is very strong. Any remaining Phusion DNA Polymerase will degrade the A overhangs, thus creating blunt ends again.

Can Phusion DNA Polymerases extend at 1 second/kb?

Yes it is possible, especially when amplifying smaller amplicons. Processivity of Phusion DNA Polymerases is 10 times that of Pfu. We recommend extension times of 15 s/kb for Phusion Flash PCR Master Mix. 15 s/kb is a conservative value that we can promise to work with almost any amplicon. In many cases, significantly shorter extension times (0-5 s/kb) can be used without compromising the yield. What separates Phusion Flash DNA Polymerase from other fast polymerases is that all steps in the PCR protocol can be shortened, including annealing and denaturation. This results in extremely fast protocols as compared with any other polymerase.

Can protocols optimized for Phusion DNA Polymerase be directly applied to Phusion Hot Start II DNA Polymerase?

Yes, protocols optimized for Phusion DNA Polymerase can be applied to Phusion Hot Start II DNA Polymerase reactions.

Do Phire and Phusion Hot Start II DNA Polymerases need a separate activation step in the PCR protocol?

No separate activation step is required since Phire and Phusion Hot Start II DNA Polymerases are inactivated by a reversibly bound, specific Affibody ligand that dissociates during initial denaturation.