Phusion High-Fidelity PCR Master Mix with HF Buffer, 100 reactions - FAQs
Phusion High-Fidelity PCR Master Mix with HF Buffer, 100 reactions - FAQs
View additional product information for Phusion High-Fidelity PCR Master Mix with HF or GC Buffer - FAQs (F531S, F532S, F531L, F532L)
1 product FAQs found
Can protocols optimized for Phusion DNA Polymerase be directly applied to Phusion Hot Start II DNA Polymerase?
Yes, protocols optimized for Phusion DNA Polymerase can be applied to Phusion Hot Start II DNA Polymerase reactions.