Canine Genotypes Panel 1.1 Kit - FAQs
Canine Genotypes Panel 1.1 Kit - FAQs
View additional product information for Canine Genotypes Panel 1.1 Kit - FAQs (F860L)
3 product FAQs found
Can additional markers be added to Thermo Scientific Animal Genotyping kits?
No, extra markers cannot be added to any of the Animal Genotyping kits. All primer mixes are optimized to work in a multiplex PCR. Addition of one or more primers would require optimization of the primer concentrations and the PCR protocol.
Are primers from Thermo Scientific Animal Genotyping kits available separately for specific loci?
We do not provide primers for specific locii separately. They come only in a multiplex format. If one or several loci did not amplify from the sample, faint signals can be a result of low DNA amounts or impurities. Some loci may be more sensitive towards purity of DNA used and may vary from sample to sample. On rare occasions, an animal can have a deletion in the marker region.
Do Thermo Scientific Animal Genotyping kits include an allelic ladder?
No, none of the genotyping kits contain an allelic ladder. Animal STR loci are highly polymorphic. Each allele has too many variants to make a universal allelic ladder.