Solución de lisis, solución de lisis de sangre completa
Invitrogen™
Solución de lisis, solución de lisis de sangre completa
Solución de lisado, solución de lisado de sangre completa
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoVolumen (métrico)
GAS010S100100 ml
GAS01025 ml
Número de catálogo GAS010S100
Precio (MXN)
-
Volumen (métrico):
100 ml
Invitrogen Cal-Lyse™ is a lysing solution formulated to enable the lysis of erythrocytes in samples of anticoagulated human peripheral blood. Invitrogen Cal-Lyse™ lysing solution is intended to be used as an aid in the enumeration of leukocytes that have been stained with monoclonal antibodies for flow cytometric analysis.
View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
Para uso diagnóstico in vitro.
Especificaciones
Cantidad100 mL
Tipo de reactivoTampón de lisis celular
Condiciones de envíoTemperatura ambiente
Volumen (métrico)100 ml
Tipo de productoReactivo
Unit SizeEach
Contenido y almacenamiento
Almacenar a temperatura ambiente.
  • Vida útil: 24 meses a partir de la fecha de fabricación
  • Preguntas frecuentes
    What reagents can I use to lyse red blood cells other than the FIX & PERM Cell Permeabilization Kit?
    Cal-Lyse premixed lysis solution (Cat. Nos. GAS010 and GAS010S100) is specifically formulated to lyse erythrocytes following monoclonal antibody staining of whole blood or bone marrow. Treatment with this reagent leads to both the lysis of red blood cells and the fixation of white cells. Treatment does not affect fluorophore-labeled antibodies bound to leukocytes, and leaves morphological scatter characteristics of leukocytes intact.
    -High-Yield Lyse (Cat. No. HYL250) is a premixed, fixative-free erythrocyte-lysis solution that can be used to help eliminate red cells from whole blood for flow cytometric analysis with minimal loss of rare blood cell populations. Using this reagent, lysis of erythrocytes is performed immediately following staining of the blood samples with fluorophore-conjugated monoclonal antibodies; there is no need for a wash step. Both are IVD reagents.
    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.