Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IL31RA Antibodies

          Abnova

          IL31RA Polyclonal Antibody, MaxPab™

          View all (15) IL31RA antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite IL31RA Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          IL31RA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IL31RA Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL31RA Antibody (H00133396-D01P) in WB

          IL31RA MaxPab rabbit polyclonal antibody. Western Blot analysis of IL31RA expression in Jurkat. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL31RA Antibody in Western Blot (WB)
          IL31RA Polyclonal Antibody, MaxPab™

          Product Details

          H00133396-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          IL31RA (ENSP00000297015, 1 a.a. approximately 732 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MMWTWALWML PSLCKFSLAA LPAKPENISC VYYYRKNLTC TWSPGKETSY TQYTVKRTYA FGEKHDNCTT NSSTSENRAS CSFFLPRITI PDNYTIEVEA ENGDGVIKSH MTYWRLENIA KTEPPKIFRV KPVLGIKRMI QIEWIKPELA PVSSDLKYTL RFRTVNSTSW MEVNFAKNRK DKNQTYNLTG LQPFTEYVIA LRCAVKESKF WSDWSQEKMG MTEEEAPCGL ELWRVLKPAE ADGRRPVRLL WKKARGAPVL EKTLGYNIWY YPESNTNLTE TMNTTNQQLE LHLGGESFWV SMISYNSLGK SPVATLRIPA IQEKSFQCIE VMQACVAEDQ LVVKWQSSAL DVNTWMIEWF PDVDSEPTTL SWESVSQATN WTIQQDKLKP FWCYNISVYP MLHDKVGEPY SIQAYAKEGV PSEGPETKVE NIGVKTVTIT WKEIPKSERK GIICNYTIFY QAEGGKGFSK TVNSSILQYG LESLKRKTSY IVQVMASTSA GGTNGTSINF KTLSFSVFEI ILITSLIGGG LLILIILTVA YGLKKPNKLT HLCWPTVPNP AESSIATWHG DDFKDKLNLK ESDDSVNTED RILKPCSTPS DKLVIDKLVV NFGNVLQEIF TDEARTGQEN NLGGEKNGYV TCPFRPDCPL GKSFEELPVS PEIPPRKSQY LRSRMPEGTR PEAKEQLLFS GQSLVPDHLC EEGAPNPYLK NSVTAREFLV SEKLPEHTKG EV

          Target Information

          IL-31 is a T cell cytokine that is preferentially produced by T helper type 2 cells. IL-31 signals through a heterodimeric receptor composed of the IL-31 receptor (IL-31R) and the oncostatin M receptor (OSM). This receptor complex recruits JAK1, JAK2, Stat1, Stat3 and Stat5 signaling pathways, as well as the PI3 kinase/AKT cascade. SHP-2 and Shc adapter molecules are also recruited and contribute to an increased activation of the MAP kinase pathway in response to IL-31. Overexpression of IL-31 in mice results in pruritus and skin dermatitis resembling human atopic dermatitis (AD). Comparisons between skin from patients with AD and healthy skin showed IL-31R expression at higher levels on epidermal keratinocytes in AD samples. Infiltrating cells, more numerous in skin from patients with AD compared with that of healthy individuals, expressed IL-31 mRNA. IL-31 may participate in the cause of itch sensation and promote scratching behavior in NC/Nga mice with atopic dermatitis, and may represent a novel target for antipruritic drug development.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: class I cytokine receptor; CRL 3; CRL3Glmr; Cytokine receptor-like 3; GLM R; GLM-R; GLM-RMGC125346; GP130 like receptor; Gp130-like monocyte receptor; Gp130-like receptor; HGLM R; hGLM-R; HGLMR; IL 31RA; IL-31 receptor subunit alpha; IL-31R subunit alpha; IL-31RAGPLCRL; IL31R subunit alpha; Interleukin-31 receptor subunit alpha; MGC125346; novel soluble type I cytokine receptor; PLCA 2; soluble type I cytokine receptor CRL3; type I cytokine receptor; unnamed protein product; ZcytoR17

          View more View less

          Gene Aliases: CRL; CRL3; GLM-R; GLMR; GPL; hGLM-R; IL-31RA; IL31RA; PLCA2; PRO21384; UNQ6368/PRO21073/PRO21384; zcytoR17

          View more View less

          UniProt ID: (Human) Q8NI17

          View more View less

          Entrez Gene ID: (Human) 133396

          View more View less

          Function(s)
          transcription coactivator activity cytokine receptor activity protein kinase binding cytokine binding
          Process(es)
          MAPK cascade glandular epithelial cell differentiation immune system process acute inflammatory response to antigenic stimulus defense response transmembrane receptor protein tyrosine kinase signaling pathway JAK-STAT cascade positive regulation of cell proliferation cytokine-mediated signaling pathway cell differentiation monocyte differentiation macrophage differentiation positive regulation of tyrosine phosphorylation of STAT protein homeostatic process negative regulation of macrophage activation negative regulation of apoptotic process positive regulation of transcription, DNA-templated receptor signaling pathway via STAT defense response to other organism
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0