HCS DNA Damage Kit - FAQs
HCS DNA Damage Kit - FAQs
View additional product information for HCS DNA Damage Kit - FAQs (H10292)
1 product FAQs found
What are the fluorescence excitation/emission maxima for the dyes contained in the HCS DNA Damage Kit?
Hoechst 33342: 350/461 nm, bound to DNA
Image-IT Dead Green viability stain: 488/515 nm
Alexa Fluor 555 goat anti-mouse IgG: 555/565 nm