What is the fluorescence excitation/emission maxima of the stains in the HCS mitochondrial health kit?
The fluorescence excitation/emission maxima are as follows: Hoechst 33342 - 350/461 nm bound to DNA; Image-iT DEAD Green viability stain - 488/515 nm; MitoHealth stain - 550/580 nm.