Product Image
Invitrogen™
Mifepristone
Mifepristone is a progesterone antagonist that activates transcription in the GeneSwitch™ System. Subphysiological levels of this steroid compound are usedRead more
Have Questions?
Catalog NumberQuantity
H11001100 μg
Catalog number H11001
Price (USD)
486.00
Each
Quantity:
100 μg
Price (USD)
486.00
Each
Mifepristone is a progesterone antagonist that activates transcription in the GeneSwitch™ System. Subphysiological levels of this steroid compound are used to induce expression from very low basal to high induced levels.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatLyophilized
Product TypeMifepristone
Quantity100 μg
Unit SizeEach
Contents & Storage
100 μg mifepristone is provided lyophilized. Store at -20°C. Mifepristone is guaranteed stable for 6 months when properly stored.
Citations & References (1)
Citations & References
Abstract
ATP-binding cassette transporter A1 mediates cellular secretion of alpha-tocopherol.
Authors: Oram J F; Vaughan A M; Stocker R;
Journal:J Biol Chem
PubMed ID:11546785
Alpha-tocopherol (alpha-TOH) is associated with plasma lipoproteins and accumulates in cell membranes throughout the body, suggesting that lipoproteins play a role in transporting alpha-TOH between tissues. Here we show that secretion of alpha-TOH from cultured cells is mediated in part by ABCA1, an ATP-binding cassette protein that transports cellular cholesterol ... More