(R)-(-)-4-Phenyl-2-oxazolidinone, 98%
(R)-(-)-4-Phenyl-2-oxazolidinone, 98%
(R)-(-)-4-Phenyl-2-oxazolidinone, 98%
Thermo Scientific Chemicals

(R)-(-)-4-Phenyl-2-oxazolidinone, 98%

CAS: 90319-52-1 | C9H9NO2 | 163.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27171.03
also known as H27171-03
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
43.20
Save 6.55 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 30-Jun-2026
43.20
Save 6.55 (15%)
Each
Chemical Identifiers
CAS90319-52-1
IUPAC Name(4R)-4-phenyl-1,3-oxazolidin-2-one
Molecular FormulaC9H9NO2
InChI KeyQDMNNMIOWVJVLY-QMMMGPOBSA-N
SMILESO=C1N[C@@H](CO1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥97.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)128.0-134.0?C
Optical Rotation-55.9 ± 0.1? (C=1 in chloroform)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only