L-m-Tyrosine, 97+%
L-m-Tyrosine, 97+%
L-m-Tyrosine, 97+%
Thermo Scientific Chemicals

L-m-Tyrosine, 97+%

CAS: 587-33-7 | C9H11NO3 | 181.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27224.MD
also known as H27224-MD
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS587-33-7
IUPAC Name(2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
Molecular FormulaC9H11NO3
InChI KeyJZKXXXDKRQWDET-SVGMAFHSNA-N
SMILESN[C@@H](CC1=CC=CC(O)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
FormPowder
Assay (HPLC)≥97.0%
Optical Rotation-9.6 ± 0.3? (C=4 in 1M HCl)
An unnatural amino acid, that has demonstrated potential in treating Parkinsons disease, Alzheimers disease, and arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in dimethyl sulfoxide, dilute acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
RUO – Research Use Only

General References:

  1. Fell V,; Greenway AM,; Hoskins JA. The metabolism of L-m-tyrosine in man.. Biochem Med. 1979, 22 (2), 246-55.
  2. Isono F,; Inukai M,; Takahashi S,; Haneishi T,; Kinoshita T,; Kuwano H. Mureidomycins A-D, novel peptidylnucleoside antibiotics with spheroplast forming activity. II. Structural elucidation. J Antibiot (Tokyo). 1989, 42 (5),667-73.