(2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate
Thermo Scientific Chemicals

(2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate

CAS: 896437-63-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27604.03
also known as H27604-03
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
285.65
Online Exclusive
317.00
Save 31.35 (10%)
Each
Specifications
CAS896437-63-1
Chemical Name or Material(2-Hydroxy-3-N,N,N-trimethylamino)propyl beta-cyclodextrin chloride hydrate
MDL NumberMFCD00078139
Melting Point∽280°C (decomposition)
RTECS NumberGU2293000
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only