Brivudine, 98%
Brivudine, 98%
Brivudine, 98%
Thermo Scientific Chemicals

Brivudine, 98%

CAS: 69304-47-8 | C11H13BrN2O5 | 333.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Catalog number H27853.03
also known as H27853-03
Price (USD)
520.65
Special offer
Online exclusive
Ends: 30-Jun-2026
612.00
Save 91.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
520.65
Special offer
Online exclusive
Ends: 30-Jun-2026
612.00
Save 91.35 (15%)
Each
Chemical Identifiers
CAS69304-47-8
IUPAC Name5-[(1E)-2-bromoethenyl]-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC11H13BrN2O5
InChI KeyODZBBRURCPAEIQ-PIXDULNESA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C=CBr)C(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder
Proton NMRConforms to structure
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥97.5%
It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in water and methanol.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. C. Desgranges.; G. Razaka.; M. Rabaud.; H. Bricaud .; J. Balzarini.; E. de Clercq. Phosphorolysis of (E)-5-(2-bromovinyl)-2'-deoxyuridine (BVDU) and other 5-substituted-2'-deoxyuridines by purified human thymidine phosphorylase and intact blood platelets.Biochem. Pharmacol. 1983, 32 (23),3583-3590 .