1-Boc-3-aminoazetidine, 94%
1-Boc-3-aminoazetidine, 94%
1-Boc-3-aminoazetidine, 94%
Thermo Scientific Chemicals

1-Boc-3-aminoazetidine, 94%

CAS: 193269-78-2 | C8H16N2O2 | 172.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H28218.03
also known as H28218-03
Price (USD)
425.65
Special offer
Online exclusive
Ends: 30-Jun-2026
501.00
Save 75.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
425.65
Special offer
Online exclusive
Ends: 30-Jun-2026
501.00
Save 75.35 (15%)
Each
Chemical Identifiers
CAS193269-78-2
IUPAC Nametert-butyl 3-aminoazetidine-1-carboxylate
Molecular FormulaC8H16N2O2
InChI KeyWPGLRFGDZJSQGI-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CC(N)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥93.0%
FormLiquid
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only