Thermo Scientific Chemicals

L-Phenylalanine benzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H28407.14
25 g, Each
Thermo Scientific Chemicals

L-Phenylalanine benzyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: H28407.14
25 g, Each
Quantity
Catalog number: H28407.14
also known as H28407-14
Price (USD)
Price: 253.00
Online price: 218.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2462-32-0
IUPAC Name
benzyl (2S)-2-amino-3-phenylpropanoate hydrochloride
Molecular Formula
C16H18ClNO2
InChI Key
CEXFHIYDTRNBJD-RSAXXLAASA-N
SMILES
Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Appearance (Color)
White
Form
Powder
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Optical Rotation
-12.5 ± 1.0? (C=1 in 4:1 acetic acid/water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text