(S)-4-Amino-2-hydroxybutyric acid, 98%
(S)-4-Amino-2-hydroxybutyric acid, 98%
(S)-4-Amino-2-hydroxybutyric acid, 98%
Thermo Scientific Chemicals

(S)-4-Amino-2-hydroxybutyric acid, 98%

CAS: 40371-51-5 | C4H9NO3 | 119.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H28796.06
also known as H28796-06
Price (USD)
61.65
Online Exclusive
68.00
Save 6.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.00
Save 6.35 (9%)
Each
Chemical Identifiers
CAS40371-51-5
IUPAC Name(2S)-4-azaniumyl-2-hydroxybutanoate
Molecular FormulaC4H9NO3
InChI KeyIVUOMFWNDGNLBJ-VKHMYHEASA-N
SMILES[NH3+]CC[C@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Optical Rotation-28? to -32? (C= 1 in water)
Water Content (Karl Fischer Titration)≤2%
FormCrystals or powder or crystalline powder or chunks
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only