2-(Boc-amino)ethanol, 95%
2-(Boc-amino)ethanol, 95%
2-(Boc-amino)ethanol, 95%
Thermo Scientific Chemicals

2-(Boc-amino)ethanol, 95%

CAS: 26690-80-2 | C7H15NO3 | 161.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32356.06
also known as H32356-06
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS26690-80-2
IUPAC Nametert-butyl N-(2-hydroxyethyl)carbamate
Molecular FormulaC7H15NO3
InChI KeyGPTXCAZYUMDUMN-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCCO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Assay (GC)≥94.0%
Refractive Index1.4450-1.4510 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Suzuki reaction. 2-(Boc-amino)ethanol as reagent in Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells.

Solubility
Soluble in water or 1% acetic acid. Soluble in ethyl acetate and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. S.F. Martin, J.A. Josey. A general protocol for the preparation of phospholipids via phosphite coupling. Tetrahedron Lett. 1988, 293631.
  2. Mamiko Ikeda,; Hidehiko Nakagawa,; Takayoshi Suzuki,; Naoki Miyata.Novel bisbenzimide-nitroxides for nuclear redox imaging in living cells. Bioorganic & Medicinal Chemistry Letters. 2012, 22(5), 1949-1952.