Fosinopril (sodium), 98%
Fosinopril (sodium), 98%
Fosinopril (sodium), 98%
Thermo Scientific™

Fosinopril (sodium), 98%

CAS: 88889-14-9 | C{3}{0}H{4}{5}NO{7}PNa, C30H45NNaO7P | 585.65 g/mol
Have Questions?
Catalog NumberQuantity
H37120.03
also known as H37120-03
1 g
Catalog number H37120.03
also known as H37120-03
Price (USD)
615.65
Online exclusive
684.00
Save 68.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
615.65
Online exclusive
684.00
Save 68.35
Each
Chemical Identifiers
CAS88889-14-9
IUPAC Namesodium (2S,4S)-4-cyclohexyl-1-(2-{[2-methyl-1-(propanoyloxy)propoxy](4-phenylbutyl)phosphoryl}acetyl)pyrrolidine-2-carboxylate
Molecular FormulaC30H45NNaO7P
InChI KeyTVTJZMHAIQQZTL-HREVRLCXSA-M
SMILES[Na+].CCC(=O)OC(OP(=O)(CCCCC1=CC=CC=C1)CC(=O)N1C[C@@H](C[C@H]1C([O-])=O)C1CCCCC1)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5% (HPLC, US-sourced material)
Mass SpecConforms (US-sourced material)
Appearance (Color)White
FormPowder
Assay (HPLC)≥97.5% (UK-sourced material)
View more