Ropivacaine, 97%
Thermo Scientific Chemicals

Ropivacaine, 97%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H37974.03
also known as H37974-03
Price (USD)
134.65
Special offer
Online exclusive
Ends: 28-Dec-2026
158.00
Save 23.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
134.65
Special offer
Online exclusive
Ends: 28-Dec-2026
158.00
Save 23.35
Each
Chemical Identifiers
CAS84057-95-4
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Melting Point (clear melt)144.0-154.0?C
Appearance (Color)White
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.