Glycine n-propyl ester hydrochloride
Glycine n-propyl ester hydrochloride
Glycine n-propyl ester hydrochloride
Thermo Scientific Chemicals

Glycine n-propyl ester hydrochloride

CAS: 13049-01-9 | C5H12ClNO2 | 153.606 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50250.03
also known as H50250-03
Price (USD)
42.40
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
42.40
Each
Specifications
CAS13049-01-9
Chemical Name or MaterialGlycine n-propyl ester hydrochloride
EINECS Number235-925-0
Formula Weight117.15
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only