(S)-3-Amino-5-hexynoic acid hydrochloride, 95%
(S)-3-Amino-5-hexynoic acid hydrochloride, 95%
(S)-3-Amino-5-hexynoic acid hydrochloride, 95%
Thermo Scientific Chemicals

(S)-3-Amino-5-hexynoic acid hydrochloride, 95%

CAS: 270596-46-8 | C6H10ClNO2 | 163.601 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52010.03
also known as H52010-03
Price (USD)
561.65
Online Exclusive
624.00
Save 62.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
561.65
Online Exclusive
624.00
Save 62.35 (10%)
Each
Specifications
CAS270596-46-8
Chemical Name or Material(S)-3-Amino-5-hexynoic acid hydrochloride
Formula Weight163.6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

Solubility
Soluble in water (90 g/L at 25°C).

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only