(S)-3-Amino-3-(3-nitrophenyl)propionic acid, 95%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid, 95%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid, 95%
Thermo Scientific Chemicals

(S)-3-Amino-3-(3-nitrophenyl)propionic acid, 95%

CAS: 734529-57-8 | C9H10N2O4 | 210.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52027.03
also known as H52027-03
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
188.65
Online Exclusive
209.00
Save 20.35 (10%)
Each
Specifications
CAS734529-57-8
Chemical Name or Material(S)-3-Amino-3-(3-nitrophenyl)propionic acid
Formula Weight210.19
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only