(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%
(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%
(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%
Thermo Scientific Chemicals

(S)-3-(Boc-amino)-4-phenylbutyric acid, 95%

CAS: 51871-62-6 | C15H21NO4 | 279.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52174.03
also known as H52174-03
Price (USD)
250.65
Special offer
Online exclusive
Ends: 30-Jun-2026
295.00
Save 44.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
250.65
Special offer
Online exclusive
Ends: 30-Jun-2026
295.00
Save 44.35 (15%)
Each
Specifications
Beilstein3060457
CAS51871-62-6
Chemical Name or Material(S)-3-(Boc-amino)-4-phenylbutyric acid
Formula Weight279.34
IUPAC Name(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only