3-(Boc-amino)cyclohexanecarboxylic acid, 98+%
3-(Boc-amino)cyclohexanecarboxylic acid, 98+%
3-(Boc-amino)cyclohexanecarboxylic acid, 98+%
Thermo Scientific Chemicals

3-(Boc-amino)cyclohexanecarboxylic acid, 98+%

CAS: 334932-13-7 | C12H21NO4 | 243.303 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52299.06
also known as H52299-06
Price (USD)
427.65
Special offer
Online exclusive
Ends: 30-Jun-2026
503.00
Save 75.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
427.65
Special offer
Online exclusive
Ends: 30-Jun-2026
503.00
Save 75.35 (15%)
Each
Specifications
CAS334932-13-7
Chemical Name or Material3-(Boc-amino)cyclohexanecarboxylic acid
Formula Weight243.3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
3-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water (2.1 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Fariborz Firooznia; Adrian Wai-Hing Cheung; John Brinkman; Joseph Grimsby; Mary Lou Gubler; Rachid Hamid; Nicholas Marcopulos; Gwendolyn Ramsey; Jenny Tan; Yang Wen; Ramakanth Sarabu. Discovery of benzothiazole-based adenosine A2B receptor antagonists with improved A2A selectivity. Bioorganic & Medicinal Chemistry Letters. 2011, 21, (7),1933-1936