(+/-)-3-(Boc-amino)pyrrolidine, 97%
(+/-)-3-(Boc-amino)pyrrolidine, 97%
(+/-)-3-(Boc-amino)pyrrolidine, 97%
Thermo Scientific Chemicals

(+/-)-3-(Boc-amino)pyrrolidine, 97%

CAS: 99724-19-3 | C9H18N2O2 | 186.255 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52414.06
also known as H52414-06
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
173.00
Save 25.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
173.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS99724-19-3
IUPAC Name(3R)-3-{[(tert-butoxy)carbonyl]amino}pyrrolidin-1-ium
Molecular FormulaC9H19N2O2
InChI KeyDQQJBEAXSOOCPG-SSDOTTSWSA-O
SMILESCC(C)(C)OC(=O)N[C@@H]1CC[NH2+]C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (GC)> 96%
Loss on Drying0.5% max
Proton NMRConforms to structure
FormCrystals or powder
View more
(±)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.

Solubility
Soluble in Methanol(almost transparency).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

General References:

  1. Mark Andrews,; Alan Brown,; Jean-Yves Chiva,; David Fradet,; David Gordon,; Mark Lansdell,; Malcolm MacKenny. Design and optimisation of selective serotonin re-uptake inhibitors with high synthetic accessibility: Part 2. Bioorganic & Medicinal Chemistry Letters. 2009, 19 (20), 5893-5897.
  2. Karine Lavrador,; Brian Murphy,; John Saunders,; Scott Struthers,; Xiaochuan Wang,; John Williams. A Screening Library for Peptide Activated G-Protein Coupled Receptors. 1. The Test Set . J. Med. Chem. 2004 , 47 (27), 6864-6874.