N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%
Thermo Scientific Chemicals

N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine, 97%

CAS: 85326-10-9 | C20H33N5O5Si | 451.599 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52726.06
also known as H52726-06
Price (USD)
647.65
Online Exclusive
719.00
Save 71.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
647.65
Online Exclusive
719.00
Save 71.35 (10%)
Each
Specifications
CAS85326-10-9
Chemical Name or MaterialN-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine
Formula Weight451.6
IUPAC NameN-[9-[(2R,4S,5R)-5-[[tert-butyl(dimethyl)silyl]oxymethyl]-4-hydroxyoxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide
InChI KeyWZQWRHWLQPEBFF-BFHYXJOUSA-N
View more
N-Isobutyryl-5'-O-tert-butyldimethylsilyl-2'-deoxyguanosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isobutyryl-5′-O-tert-butyldimethylsilyl-2′-deoxyguanosine is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Store away from oxidizing agents.
RUO – Research Use Only